Assay Details
Gene Symbol: MCHR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503901.3 | XP_001503951.1 | 578 | 99 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
