Assay Details
Gene Symbol: HARS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504153.6 | XP_001504203.1 | 909 | 62 | ||
XM_014730619.2 | XP_014586105.1 | 835 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
