Assay Details
Gene Symbol: IL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504354.2 | XP_001504404.1 | 114 | 98 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
