Assay Details
Gene Symbol: CFAP73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001915005.3 | XP_001915040.2 | 836 | 56 | ||
XM_023647119.1 | XP_023502887.1 | 842 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
