Assay Details
Gene Symbol: DPEP2NB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014738501.2 | XP_014593987.1 | 1007 | 125 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014738501.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
