Assay Details
Gene Symbol: TNFRSF13C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001245977.1 | NP_001232906.1 | 1 - 2 | 129 | 102 | |
GenBank mRNA | JF502272.1 | - | 1 - 2 | 129 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
