Assay Details
Gene Symbol: ECM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015293485.2 | XP_015148971.1 | 3 - 4 | 622 | 101 | |
XM_414297.6 | XP_414297.2 | 3 - 4 | 878 | 101 | ||
GenBank mRNA | BX929391.1 | - | 3 - 4 | 588 | 101 | |
BX933407.2 | - | 1 - 2 | 93 | 101 | ||
CR353762.1 | - | 4 - 5 | 624 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
