Assay Details
Gene Symbol: SERHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277373.1 | NP_001264302.1 | 2 - 3 | 297 | 80 | |
XM_015285070.2 | XP_015140556.1 | 2 - 3 | 261 | 80 | ||
XM_025148464.1 | XP_025004232.1 | 2 - 3 | 408 | 80 | ||
GenBank mRNA | BX929989.1 | - | 1 - 2 | 204 | 80 | |
BX932388.1 | - | 1 - 2 | 184 | 80 | ||
CD218551.1 | - | 2 - 3 | 266 | 80 | ||
CD735806.1 | - | 2 - 3 | 297 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
