Assay Details
Gene Symbol: SERHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277373.1 | NP_001264302.1 | 4 - 5 | 454 | 92 | |
XM_025148464.1 | XP_025004232.1 | 4 - 5 | 565 | 92 | ||
GenBank mRNA | BX929989.1 | - | 3 - 4 | 361 | 92 | |
BX932388.1 | - | 3 - 4 | 341 | 92 | ||
CD218551.1 | - | 4 - 5 | 423 | 92 | ||
DN929581.1 | - | 2 - 3 | 80 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
