Assay Details
Gene Symbol: MRPL22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025154991.1 | XP_025010759.1 | 2 - 3 | 114 | 79 | |
XM_025154992.1 | XP_025010760.1 | 2 - 3 | 114 | 79 | ||
XM_414573.6 | XP_414573.1 | 2 - 3 | 114 | 79 | ||
GenBank mRNA | BX930154.1 | - | 2 - 3 | 72 | 79 | |
BX934781.1 | - | 2 - 3 | 83 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
