Assay Details
Gene Symbol: IRAK1BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278022.1 | NP_001264951.1 | 2 - 3 | 392 | 154 | |
GenBank mRNA | BX930608.1 | - | 2 - 3 | 392 | 154 |

More Information
Set Membership: |
![]() ![]() ![]() |
