Assay Details
Gene Symbol: RPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_418679.5 | XP_418679.1 | 2 - 3 | 229 | 80 | |
GenBank mRNA | BX931405.1 | - | 2 - 3 | 176 | 80 | |
BX934509.1 | - | 2 - 3 | 198 | 80 | ||
BX936195.2 | - | 2 - 3 | 217 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
