Assay Details
Gene Symbol: DPCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277365.1 | NP_001264294.1 | 2 - 3 | 187 | 73 | |
NM_001277366.1 | NP_001264295.1 | 2 - 3 | 169 | 73 | ||
GenBank mRNA | BU399154.1 | - | 2 - 3 | 156 | 73 | |
BX931687.2 | - | 1 - 2 | 406 | 73 | ||
BX931759.1 | - | 2 - 3 | 164 | 73 | ||
BX932644.1 | - | 2 - 3 | 162 | 73 | ||
BX932782.1 | - | 2 - 3 | 179 | 73 | ||
BX933238.1 | - | 2 - 3 | 163 | 73 | ||
CR386779.1 | - | 2 - 3 | 145 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
