Assay Details
Gene Symbol: SDR42E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004944259.3 | XP_004944316.3 | 1 - 2 | 45 | 49 | |
XM_004944261.3 | XP_004944318.3 | 1 - 2 | 37 | 49 | ||
XM_414167.6 | XP_414167.4 | 1 - 2 | 31 | 49 | ||
GenBank mRNA | BX931784.1 | - | 1 - 2 | 34 | 49 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004944259.3, XM_004944261.3, XM_414167.6 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
