Assay Details
Gene Symbol: TPPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001231863.5 | XP_001231864.1 | 3 - 4 | 555 | 155 | |
XM_004935074.3 | XP_004935131.3 | 4 - 5 | 778 | 155 | ||
XM_015275922.2 | XP_015131408.1 | 3 - 4 | 544 | 155 | ||
XM_025147612.1 | XP_025003380.1 | 3 - 4 | 519 | 155 | ||
XM_418894.6 | XP_418894.3 | 4 - 5 | 660 | 155 | ||
GenBank mRNA | BX932523.2 | - | 4 - 5 | 625 | 155 | |
BX932684.1 | - | 4 - 5 | 645 | 155 | ||
BX934333.2 | - | 4 - 5 | 649 | 155 | ||
BX935488.2 | - | 3 - 4 | 523 | 155 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
