Assay Details
Gene Symbol: RPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277164.1 | NP_001264093.1 | 2 - 3 | 178 | 114 | |
GenBank mRNA | BX933609.1 | - | 1 - 2 | 164 | 114 | |
BX934117.1 | - | 1 - 2 | 161 | 114 | ||
BX934597.2 | - | 1 - 2 | 164 | 114 | ||
BX935640.2 | - | 1 - 2 | 164 | 114 | ||
CN222435.1 | - | 2 - 3 | 179 | 114 | ||
CN226583.1 | - | 1 - 2 | 169 | 114 | ||
CR386179.1 | - | 1 - 2 | 68 | 114 | ||
CR386183.1 | - | 1 - 2 | 160 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
