Assay Details
Gene Symbol: ATP5PB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_417993.5 | XP_417993.3 | 312 | 86 | ||
GenBank mRNA | BX932164.2 | - | 282 | 86 | ||
BX934049.1 | - | 299 | 86 | |||
BX934066.1 | - | 311 | 86 | |||
CR406487.1 | - | 284 | 86 | |||
CR407330.1 | - | 292 | 86 | |||
CR407332.1 | - | 285 | 86 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
