Assay Details
Gene Symbol: ATP5PB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_417993.5 | XP_417993.3 | 595 | 72 | ||
GenBank mRNA | BX932164.2 | - | 505 | 72 | ||
BX934049.1 | - | 582 | 72 | |||
BX934066.1 | - | 594 | 72 | |||
CR406487.1 | - | 567 | 72 | |||
CR407330.1 | - | 575 | 72 | |||
CR407332.1 | - | 568 | 72 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
