Assay Details
Gene Symbol: CYB561A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199574.1 | NP_001186503.1 | 3 - 4 | 539 | 150 | |
XM_015286081.2 | XP_015141567.1 | 3 - 4 | 584 | 150 | ||
XM_015286082.2 | XP_015141568.1 | 3 - 4 | 449 | 150 | ||
XM_015286083.2 | XP_015141569.1 | 3 - 4 | 529 | 150 | ||
GenBank mRNA | BX930104.2 | - | 2 - 3 | 429 | 150 | |
BX934774.2 | - | 3 - 4 | 489 | 150 | ||
BX934944.1 | - | 2 - 3 | 423 | 150 | ||
BX950333.1 | - | 3 - 4 | 539 | 150 | ||
CD216049.1 | - | 3 - 4 | 471 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
