Assay Details
Gene Symbol: EPFIP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025149944.1 | XP_025005712.1 | 5 - 6 | 688 | 116 | |
XM_025149945.1 | XP_025005713.1 | 5 - 6 | 535 | 116 | ||
GenBank mRNA | BX932745.1 | - | 5 - 6 | 383 | 116 | |
BX935742.1 | - | 4 - 5 | 288 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
