Assay Details
Gene Symbol: SMIM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197056.1 | NP_001183985.1 | 2 - 3 | 384 | 86 | |
NM_001282053.1 | NP_001268982.1 | 2 - 3 | 374 | 86 | ||
GenBank mRNA | BX931964.2 | - | 1 - 2 | 499 | 86 | |
BX932456.1 | - | 2 - 3 | 315 | 86 | ||
BX935345.1 | - | 2 - 3 | 260 | 86 | ||
BX936112.1 | - | 2 - 3 | 308 | 86 | ||
CR354283.1 | - | 2 - 3 | 293 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
