Assay Details
Gene Symbol: LLGL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015295434.2 | XP_015150920.1 | 3 - 4 | 250 | 134 | |
XM_015295435.2 | XP_015150921.1 | 3 - 4 | 250 | 134 | ||
XM_015295436.2 | XP_015150922.1 | 5 - 6 | 909 | 134 | ||
XM_015295438.2 | XP_015150924.1 | 4 - 5 | 862 | 134 | ||
XM_015295439.2 | XP_015150925.1 | 4 - 5 | 824 | 134 | ||
XM_025141654.1 | XP_024997422.1 | 3 - 4 | 250 | 134 | ||
XM_025141655.1 | XP_024997423.1 | 3 - 4 | 474 | 134 | ||
XM_025141656.1 | XP_024997424.1 | 4 - 5 | 373 | 134 | ||
XM_025141657.1 | XP_024997425.1 | 4 - 5 | 811 | 134 | ||
XM_025141658.1 | XP_024997426.1 | 3 - 4 | 764 | 134 | ||
GenBank mRNA | BX930166.2 | - | 3 - 4 | 479 | 134 | |
BX930538.2 | - | 3 - 4 | 257 | 134 | ||
BX931931.2 | - | 3 - 4 | 267 | 134 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015295439.2, XM_025141657.1, XM_025141658.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
