Assay Details
Gene Symbol: LLGL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015295434.2 | XP_015150920.1 | 4 - 5 | 320 | 115 | |
XM_015295435.2 | XP_015150921.1 | 4 - 5 | 320 | 115 | ||
XM_015295436.2 | XP_015150922.1 | 6 - 7 | 979 | 115 | ||
XM_015295438.2 | XP_015150924.1 | 5 - 6 | 932 | 115 | ||
XM_015295439.2 | XP_015150925.1 | 5 - 6 | 894 | 115 | ||
XM_025141654.1 | XP_024997422.1 | 4 - 5 | 320 | 115 | ||
XM_025141655.1 | XP_024997423.1 | 4 - 5 | 544 | 115 | ||
XM_025141656.1 | XP_024997424.1 | 5 - 6 | 443 | 115 | ||
XM_025141657.1 | XP_024997425.1 | 5 - 6 | 881 | 115 | ||
XM_025141658.1 | XP_024997426.1 | 4 - 5 | 834 | 115 | ||
GenBank mRNA | BX930166.2 | - | 4 - 5 | 549 | 115 | |
BX930538.2 | - | 4 - 5 | 327 | 115 | ||
BX931931.2 | - | 4 - 5 | 337 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
