Assay Details
Gene Symbol: LLGL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015295434.2 | XP_015150920.1 | 5 - 6 | 442 | 72 | |
XM_015295435.2 | XP_015150921.1 | 5 - 6 | 442 | 72 | ||
XM_015295436.2 | XP_015150922.1 | 7 - 8 | 1101 | 72 | ||
XM_015295438.2 | XP_015150924.1 | 6 - 7 | 1054 | 72 | ||
XM_015295439.2 | XP_015150925.1 | 6 - 7 | 1016 | 72 | ||
XM_025141654.1 | XP_024997422.1 | 5 - 6 | 442 | 72 | ||
XM_025141655.1 | XP_024997423.1 | 5 - 6 | 666 | 72 | ||
XM_025141656.1 | XP_024997424.1 | 6 - 7 | 565 | 72 | ||
XM_025141657.1 | XP_024997425.1 | 6 - 7 | 1003 | 72 | ||
XM_025141658.1 | XP_024997426.1 | 5 - 6 | 956 | 72 | ||
GenBank mRNA | BX930166.2 | - | 5 - 6 | 671 | 72 | |
BX930538.2 | - | 5 - 6 | 449 | 72 | ||
BX931931.2 | - | 5 - 6 | 459 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
