Assay Details
Gene Symbol: LLGL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015295434.2 | XP_015150920.1 | 6 - 7 | 605 | 84 | |
XM_015295435.2 | XP_015150921.1 | 6 - 7 | 605 | 84 | ||
XM_015295436.2 | XP_015150922.1 | 8 - 9 | 1264 | 84 | ||
XM_015295438.2 | XP_015150924.1 | 7 - 8 | 1217 | 84 | ||
XM_015295439.2 | XP_015150925.1 | 7 - 8 | 1179 | 84 | ||
XM_025141654.1 | XP_024997422.1 | 6 - 7 | 605 | 84 | ||
XM_025141655.1 | XP_024997423.1 | 6 - 7 | 829 | 84 | ||
XM_025141656.1 | XP_024997424.1 | 7 - 8 | 728 | 84 | ||
XM_025141657.1 | XP_024997425.1 | 7 - 8 | 1166 | 84 | ||
XM_025141658.1 | XP_024997426.1 | 6 - 7 | 1119 | 84 | ||
GenBank mRNA | BX930166.2 | - | 6 - 7 | 834 | 84 | |
BX930538.2 | - | 6 - 7 | 612 | 84 | ||
BX931931.2 | - | 6 - 7 | 622 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
