Assay Details
Gene Symbol: RAMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX932709.1 | - | 1 - 2 | 111 | 101 | |
BX935935.2 | - | 1 - 2 | 260 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
