Assay Details
Gene Symbol: TTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199392.2 | NP_001186321.1 | 2 - 3 | 292 | 69 | |
GenBank mRNA | BX950343.2 | - | 2 - 3 | 293 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
