Assay Details
Gene Symbol: B4GALT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025143616.1 | XP_024999384.1 | 6 - 7 | 1505 | 62 | |
XM_416563.5 | XP_416563.1 | 6 - 7 | 1136 | 62 | ||
GenBank mRNA | CR338987.2 | - | 6 - 7 | 1141 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
