Assay Details
Gene Symbol: NME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204761.1 | NP_001191690.1 | 4 | 1668 | 74 | |
XM_015295390.1 | XP_015150876.1 | 5 | 1588 | 74 | ||
XM_025141609.1 | XP_024997377.1 | 5 | 1563 | 74 | ||
XM_025141610.1 | XP_024997378.1 | 5 | 1574 | 74 | ||
GenBank mRNA | BU294775.1 | - | 1 | 391 | 74 | |
CR385461.1 | - | 1 | 851 | 74 |
Gene Symbol: LOC112530034 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025141797.1 | XP_024997565.1 | 1 | 511 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025141797.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
