Assay Details
Gene Symbol: CD24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277735.1 | NP_001264664.1 | 1 - 2 | 152 | 100 | |
GenBank mRNA | AJ445890.1 | - | 1 - 2 | 153 | 100 | |
CR385611.1 | - | 1 - 2 | 145 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
