Assay Details
Gene Symbol: IRAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001348560.1 | NP_001335489.1 | 2 - 3 | 710 | 109 | |
GenBank mRNA | CR388830.1 | - | 2 - 3 | 164 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
