Assay Details
Gene Symbol: IRAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001348560.1 | NP_001335489.1 | 3 - 4 | 890 | 76 | |
GenBank mRNA | CR388830.1 | - | 3 - 4 | 344 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
