Assay Details
Gene Symbol: LSM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277355.1 | NP_001264284.1 | 2 - 3 | 251 | 120 | |
XM_025147609.1 | XP_025003377.1 | 2 - 3 | 197 | 120 | ||
GenBank mRNA | BX932416.1 | - | 2 - 3 | 140 | 120 | |
CR390408.1 | - | 2 - 3 | 252 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
