Assay Details
Gene Symbol: LSM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277355.1 | NP_001264284.1 | 3 - 4 | 382 | 107 | |
GenBank mRNA | CR390408.1 | - | 3 - 4 | 383 | 107 | |
CR524002.1 | - | 2 - 3 | 677 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
