Assay Details
Gene Symbol: INTS6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015278456.2 | XP_015133942.2 | 21 - 22 | 6126 | 130 | |
XM_025149816.1 | XP_025005584.1 | 22 - 23 | 10569 | 130 | ||
XM_025149817.1 | XP_025005585.1 | 21 - 22 | 3957 | 130 | ||
XM_025149818.1 | XP_025005586.1 | 20 - 21 | 6520 | 130 | ||
XM_025149819.1 | XP_025005587.1 | 19 - 20 | 6387 | 130 | ||
GenBank mRNA | CR338718.2 | - | 3 - 4 | 541 | 130 | |
CR523792.1 | - | 2 - 3 | 486 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
