Assay Details
Gene Symbol: PTGES3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328354.1 | NP_001315283.1 | 4 - 5 | 369 | 70 | |
XM_025143976.1 | XP_024999744.1 | 4 - 5 | 389 | 70 | ||
GenBank mRNA | BX931852.2 | - | 4 - 5 | 342 | 70 | |
BX933009.1 | - | 4 - 5 | 344 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
