Assay Details
Gene Symbol: PTGES3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328354.1 | NP_001315283.1 | 6 - 7 | 518 | 80 | |
XM_015299632.2 | XP_015155118.1 | 6 - 7 | 579 | 80 | ||
XM_025143976.1 | XP_024999744.1 | 6 - 7 | 538 | 80 | ||
GenBank mRNA | BX931852.2 | - | 6 - 7 | 491 | 80 | |
BX933009.1 | - | 6 - 7 | 493 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
