Assay Details
Gene Symbol: DHRSX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015277734.2 | XP_015133220.1 | 5 - 6 | 706 | 67 | |
GenBank mRNA | BX931965.2 | - | 5 - 6 | 623 | 67 | |
BX934631.1 | - | 5 - 6 | 601 | 67 | ||
BX950546.2 | - | 5 - 6 | 1012 | 67 | ||
CR405833.1 | - | 4 - 5 | 491 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
