Assay Details
Gene Symbol: SLF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004949396.3 | XP_004949453.1 | 14 - 15 | 2101 | 72 | |
XM_004949397.3 | XP_004949454.1 | 14 - 15 | 2098 | 72 | ||
XM_004949398.3 | XP_004949455.1 | 14 - 15 | 2098 | 72 | ||
XM_004949399.3 | XP_004949456.1 | 13 - 14 | 1984 | 72 | ||
XM_424703.4 | XP_424703.4 | 14 - 15 | 2100 | 72 | ||
GenBank mRNA | CR407078.1 | - | 1 - 2 | 180 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
