Assay Details
Gene Symbol: MRPS35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278091.1 | NP_001265020.1 | 4 - 5 | 506 | 67 | |
NM_001278092.1 | NP_001265021.1 | 4 - 5 | 448 | 67 | ||
GenBank mRNA | BX935160.1 | - | 4 - 5 | 440 | 67 | |
CR522886.1 | - | 4 - 5 | 338 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
