Assay Details
Gene Symbol: ALKBH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277496.1 | NP_001264425.1 | 2 - 3 | 534 | 130 | |
NM_001277497.1 | NP_001264426.1 | 2 - 3 | 130 | |||
NM_001277500.1 | NP_001264429.1 | 3 - 4 | 435 | 130 | ||
GenBank mRNA | BU400522.1 | - | 2 - 3 | 130 | ||
BX931159.2 | - | 2 - 3 | 534 | 130 | ||
BX931177.1 | - | 3 - 4 | 435 | 130 | ||
CR405960.1 | - | 3 - 4 | 435 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
