Assay Details
Gene Symbol: LRRC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003642283.4 | XP_003642331.1 | 1 - 2 | 91 | 91 | |
GenBank mRNA | BX932126.1 | - | 1 - 2 | 59 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003642283.4 transcripts.
Set Membership: |
![]() ![]() ![]() |
