Assay Details
Gene Symbol: LRRC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003642283.4 | XP_003642331.1 | 2 - 3 | 217 | 75 | |
XM_015279567.2 | XP_015135053.1 | 2 - 3 | 308 | 75 | ||
XM_015279568.2 | XP_015135054.1 | 2 - 3 | 230 | 75 | ||
GenBank mRNA | BX932126.1 | - | 2 - 3 | 185 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003642283.4, XM_015279567.2, XM_015279568.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
