Assay Details
Gene Symbol: LOC422609 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015276476.2 | XP_015131962.1 | 7 - 8 | 1036 | 108 | |
XM_015276477.2 | XP_015131963.1 | 9 - 10 | 1042 | 108 | ||
XM_015276478.2 | XP_015131964.1 | 8 - 9 | 6394 | 108 | ||
XM_025150504.1 | XP_025006272.1 | 9 - 10 | 965 | 108 | ||
XM_025150505.1 | XP_025006273.1 | 7 - 8 | 1382 | 108 | ||
XM_420568.5 | XP_420568.2 | 8 - 9 | 1110 | 108 | ||
GenBank mRNA | BX932483.1 | - | 6 - 7 | 640 | 108 | |
BX934425.2 | - | 4 - 5 | 361 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
