Assay Details
Gene Symbol: EPFIP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025149944.1 | XP_025005712.1 | 3 - 4 | 520 | 121 | |
XM_025149945.1 | XP_025005713.1 | 3 - 4 | 367 | 121 | ||
GenBank mRNA | BX932745.1 | - | 3 - 4 | 215 | 121 | |
BX935742.1 | - | 2 - 3 | 120 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() |
