Assay Details
Gene Symbol: CREBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278110.3 | NP_001265039.1 | 1 - 2 | 131 | 100 | |
GenBank mRNA | BU339707.1 | - | 1 - 2 | 132 | 100 | |
BX932597.1 | - | 1 - 2 | 99 | 100 | ||
BX933150.1 | - | 1 - 2 | 98 | 100 | ||
BX933493.2 | - | 1 - 2 | 120 | 100 | ||
BX950505.1 | - | 1 - 2 | 113 | 100 | ||
CR386358.1 | - | 1 - 2 | 117 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001278110.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
