Assay Details
Gene Symbol: DCAF4L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004941670.3 | XP_004941727.1 | 1 - 2 | 44 | 77 | |
GenBank mRNA | BX933708.1 | - | 1 - 2 | 62 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004941670.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
