Assay Details
Gene Symbol: ELOVL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197309.1 | NP_001184238.1 | 2 - 3 | 162 | 76 | |
XM_015284630.2 | XP_015140116.1 | 1 - 2 | 289 | 76 | ||
GenBank mRNA | BX933731.1 | - | 1 - 2 | 131 | 76 | |
BX934330.1 | - | 1 - 2 | 131 | 76 | ||
HQ264101.1 | - | 2 - 3 | 162 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
