Assay Details
Gene Symbol: CTSV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168009.1 | NP_001161481.1 | 7 - 8 | 979 | 132 | |
GenBank mRNA | BX932251.2 | - | 7 - 8 | 965 | 132 | |
BX933970.1 | - | 7 - 8 | 966 | 132 | ||
BX936191.1 | - | 7 - 8 | 953 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
