Assay Details
Gene Symbol: PAPOLB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198601.3 | NP_001185530.1 | 6 - 7 | 761 | 120 | |
XM_015287288.2 | XP_015142774.1 | 6 - 7 | 768 | 120 | ||
XM_015287290.2 | XP_015142776.1 | 6 - 7 | 768 | 120 | ||
XM_015287291.2 | XP_015142777.1 | 6 - 7 | 768 | 120 | ||
GenBank mRNA | AF061758.1 | - | 2 - 3 | 134 | 120 | |
BX934124.1 | - | 6 - 7 | 533 | 120 | ||
BX934578.2 | - | 5 - 6 | 583 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() |
